Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Ion Standard, 0.50 ppm Na, Reagents

Catalog No. 01333915
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
01-333-915 500 mL
1 options

Catalog No. 01-333-915

Supplier: Reagents Holdings Llc CS022850500A

Only null left
Add to Cart
Add to Cart

Sodium Ion Standard, 0.50 ppm Na, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Ion Standard, 0.50 ppm Na
Packaging Poly Bottle
CAS 7732-18-5
Specific Gravity 1.0001
Physical Form Liquid
Molecular Formula NaCl/H2O
Color Colorless
pH 7
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Water Analysis
Quantity 500 mL
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.