Alfa Aesar™ Sodium isopropoxide isopropanol adduct, 95%

Manufacturer: Alfa Aesar™ 4564814

Quantity 25g

 View more versions of this product

Catalog No. AA4564814

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula NaOC3H7·C3H8O
    Linear Formula NaOCH(CH3)2·C3H7OH
    Formula Weight 142.17
    MDL Number MFCD12547728
    UN Number UN3206
    Chemical Name or Material Sodium isopropoxide isopropanol adduct

    Specifications

    Formula Weight 142.17
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula NaOC3H7·C3H8O
    Linear Formula NaOCH(CH3)2·C3H7OH
    Health Hazard 1 Danger
    Sensitivity Air & Moisture Sensitive
    Physical Form Powder
    MDL Number MFCD12547728
    UN Number UN3206
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H228-H251-H314-H318
    Flammable solid.
    Self-heating: may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Color Cream
    Melting Point 150°C (dec.)
    DOT Information Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.
    Chemical Name or Material Sodium isopropoxide isopropanol adduct
    Assay 95%

    GHS H Statement
    H228-H251-H314-H318
    Flammable solid.
    Self-heating: may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit