Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium L-Lactate, Approx. 98%, Spectrum™

Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Amber Glass Bottle
867-56-1
C3H5NaO3
112.06
NGSFWBMYFKHRBD-UHFFFAOYNA-M
sodium 2-hydroxypropanoate
[Na+].CC(O)C([O-])=O
867-56-1
C3H5NaO3
112.06
NGSFWBMYFKHRBD-UHFFFAOYNA-M
sodium 2-hydroxypropanoate
[Na+].CC(O)C([O-])=O

S2008, 867-56-1, C3H5NaO3

Spectrum™ Sodium L-Lactate , also known as milk acid, is used in various biochemical processes. It is a carboxylic acid and has a hydroxyl group adjacent to the carboxyl group. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Grade Reagent
CAS Max % 1
Percent Purity Approx. 98%
Packaging Amber Glass Bottle
Quantity 50 g