Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium L-Lactate Solution, USP, 60%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
867-56-1,7732-18-5
C3H5NaO3
112.06
NGSFWBMYFKHRBD-UHFFFAOYNA-M
sodium 2-hydroxypropanoate
[Na+].CC(O)C([O-])=O
867-56-1,7732-18-5
C3H5NaO3
112.06
NGSFWBMYFKHRBD-UHFFFAOYNA-M
sodium 2-hydroxypropanoate
[Na+].CC(O)C([O-])=O

S1324, C3H5NaO3

Spectrum™ Sodium L-Lactate Solution, 60 Percent, USP is used for replacing fluids and electrolytes in those who have low blood volume or low blood pressure. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Drum
pH 5.0 to 9.0
Quantity 20 L
CAS Max % 0.6,0.4
Percent Purity 58.8 to 61.2%
Grade USP