Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Lauryl Sulfate, Ultrapure, 98%, Spectrum™

Click to view available options
Quantity:
100 g
1 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
151-21-3
C12H25NaO4S
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
151-21-3
C12H25NaO4S
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

S1331, 151-21-3, C12H25O4SNa

Spectrum™ Sodium Lauryl Sulfate, Ultrapure , is an organic compound, and anionic surfactant derived from coconut or palm oils, for use as a detergent. As an Organosulfate, this salt has the amphiphilic properties needed in a detergent. It's uses vary from shark repellent, fruit fly killer and biocide.

CAS Max % 1
Packaging Poly Bottle
Percent Purity 98%
Grade Ultrapure
Quantity 1 kg