Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium meta-Periodate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle

Chemical Identifiers

7790-28-5
INaO4
213.89
JQWHASGSAFIOCM-UHFFFAOYSA-M
23667635
CHEBI:75226

High quality chemicals for laboratory and specialized industrial use

CAS 7790-28-5
Molecular Formula INaO4
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
SMILES [O-]I(=O)(=O)=O.[Na+]
IUPAC Name sodium;periodate
Molecular Weight (g/mol) 213.89
PubChem CID 23667635
ChEBI CHEBI:75226
Formula Weight 213.89
Packaging Poly Bottle
Specific Gravity 1L = 3.87kg
Quantity 125g
Chemical Name or Material Sodium meta-Periodate
Grade Baker Analyzed™ ACS Reagent, Meets ACS reagent chemical requirements
Show More Show Less