Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Metabisulfite, Crystalline Powder, NF, EP, BP, JP, 65-67.4%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7681-57-4
Na2O5S2
190.09
LDTLADDKFLAYJA-UHFFFAOYSA-L
disodium (sulfinatooxy)sulfinate
[Na+].[Na+].[O-]S(=O)OS([O-])=O
7681-57-4
Na2O5S2
190.09
LDTLADDKFLAYJA-UHFFFAOYSA-L
disodium (sulfinatooxy)sulfinate
[Na+].[Na+].[O-]S(=O)OS([O-])=O

S1924, 7681-57-4, Na2S2O5

Spectrum™ Sodium Metabisulfite, Crystalline Powder, NF, EP, BP, JP
Grade Multi Compendia
CAS Max % 1
Percent Purity 65 to 67.4%
Iron (Fe) 20 ppm
Packaging Poly Bottle
Quantity 500 g