Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Metabisulfite (Laboratory), Fisher Chemical

Click to view available options
Quantity:
10kg
Packaging:
Poly Pail
7681-57-4
Na2O5S2
190.095
MFCD00167602
HRZFUMHJMZEROT-UHFFFAOYSA-L
sodium metabisulfite,sodium pyrosulfite,sodium disulfite,disodium pyrosulfite,sodium metabisulphite,disodium disulfite,disodium disulphite,fertisilo,disodium metabisulfite,natrii disulfis
656671
[O-]S(=O)S(=O)(=O)[O-].[Na+].[Na+]
7681-57-4
Na2O5S2
190.095
MFCD00167602
HRZFUMHJMZEROT-UHFFFAOYSA-L
sodium metabisulfite,sodium pyrosulfite,sodium disulfite,disodium pyrosulfite,sodium metabisulphite,disodium disulfite,disodium disulphite,fertisilo,disodium metabisulfite,natrii disulfis
656671
[O-]S(=O)S(=O)(=O)[O-].[Na+].[Na+]
Formula Weight 190.11
Physical Form Powder/Solid
Grade Laboratory
Density 1.4g/cm³
Identification Pass Test
Packaging Poly Pail
Color White
Melting Point 150°C
pH 4 to 6
Quantity 10kg
Chemical Name or Material Sodium Metabisulfite
Show More Show Less

WARNING!

Emergency Overview
Harmful if swallowed. May cause skin and respiratory tract irritation. Contact with acids liberates toxic gas. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:0
Instability:1