Learn More
Sodium metaborate tetrahydrate, 98.5%, extra pure, ACROS Organics™
Specifications
Assay Percent Range | 98.5% |
Linear Formula | NaBO2·4H2O |
Formula Weight | 137.86 |
Physical Form | Crystals or Crystalline Powder |
Percent Purity | ≥98.0% |
Grade | Extra Pure |
Packaging | Glass Bottle |
Color | White |
Quantity | 25g |
Chemical Name or Material | Sodium metaborate tetrahydrate, p.a. |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility. Suspected of damaging the unborn child.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Obtain special instructions before use.
IF exposed or concerned: Get medical attention/advice.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.