Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium metaborate tetrahydrate, 98.5%, extra pure, ACROS Organics™
Click to view available options
Quantity:
25g
500g
2.5kg
10kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Specifications
Assay Percent Range | 98.5% |
Linear Formula | NaBO2·4H2O |
Merck Index | 10,8477 |
Solubility Information | Solubility in water: 41.9% (20°C) |
Formula Weight | 137.86 |
Physical Form | Crystals or Crystalline Powder |
Percent Purity | 98.5% |
Grade | Extra Pure |
Packaging | Plastic bottle |
Solubility | Solubility in water: 41.9% (20°C). |
Show More |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Suspected of damaging the unborn child.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical
GHS Signal Word: Warning