Sodium metaperiodate, ACS, 99.8-100.3%, Alfa Aesar™

Maximum level of impurities: Other halogens (as Cl) 0.02%, Mn 3ppm

Manufacturer: Alfa Aesar 4430936

Quantity 500g

 View more versions of this product

Catalog No. AA4430936

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7790-28-5
    Physical Form Powder
    Assay Percent Range 99.8-100.3%
    Molecular Formula INaO4
    Formula Weight 213.89
    MDL Number MFCD00003534
    Synonym Sodium periodate
    Chemical Name or Material Sodium metaperiodate, For analysis ACS
    Grade ACS Grade

    Specifications

    Formula Weight 213.89
    Physical Form Powder
    Quantity 500g
    Color White
    DOT Information DOT Class: 5.1, Packaging Group: II
    Density 3.86g/cm3 at 20°C
    EINECS Number 232-197-6
    Assay Percent Range 99.8-100.3%
    Odor Odorless
    Molecular Formula INaO4
    MDL Number MFCD00003534
    CAS 7790-28-5
    Synonym Sodium periodate
    Health Hazard 1 H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Sodium metaperiodate, For analysis ACS
    Grade ACS Grade

    H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit