Sodium metatungstate monohydrate, Alfa Aesar™

Manufacturer: Alfa Aesar 2253430

Quantity 250g

 View more versions of this product

Catalog No. AA2253430

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12141-67-2
    Physical Form Powder
    Molecular Formula Na6O39W12·H2O
    Formula Weight 2986.13; 2968.12 (anhydrous)
    MDL Number MFCD00061466
    Synonym Sodium polytungstate
    Chemical Name or Material Sodium metatungstate monohydrate

    Specifications

    Physical Form Powder
    Formula Weight 2986.13; 2968.12 (anhydrous)
    Quantity 250g
    Color White
    Density 5.47g/cm3
    Odor Odorless
    Molecular Formula Na6O39W12·H2O
    Appearance White powder
    MDL Number MFCD00061466
    Synonym Sodium polytungstate
    Health Hazard 1 H318-H302-H402-H412: Causes serious eye damage. Harmful if swallowed. Harmful to aquatic life. Harmful to aquatic life with long lasting effects.
    CAS 12141-67-2
    Chemical Name or Material Sodium metatungstate monohydrate

    H318-H302-H402-H412: Causes serious eye damage. Harmful if swallowed. Harmful to aquatic life. Harmful to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit