Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium methanesulfonate, 98%

Click to view available options
Quantity:
5g
25g
2386-57-4
CH3NaO3S
118.082
MFCD00064389
KKVTYAVXTDIPAP-UHFFFAOYSA-M
sodium methanesulfonate,methanesulfonic acid sodium salt,methanesulfonic acid, sodium salt,sodium methanesulphonate,sodium mesylate,methylsulfonic acid sodium salt,75-75-2 parent,sodium methyl sulfonate,methanesulfonic acid, sodium salt 1:1,sodium methylsulfonate
638112
sodium;methanesulfonate
CS(=O)(=O)[O-].[Na+]
2386-57-4
CH3NaO3S
118.082
MFCD00064389
KKVTYAVXTDIPAP-UHFFFAOYSA-M
sodium methanesulfonate,methanesulfonic acid sodium salt,methanesulfonic acid, sodium salt,sodium methanesulphonate,sodium mesylate,methylsulfonic acid sodium salt,75-75-2 parent,sodium methyl sulfonate,methanesulfonic acid, sodium salt 1:1,sodium methylsulfonate
638112
sodium;methanesulfonate
CS(=O)(=O)[O-].[Na+]
Applications
Sodium methanesulfonate is used as a non-steroidal anti-inflammatory drug.

Solubility
Soluble in water, alcohol, ether, hexane, benzene, methylcyclopentane, toluene, o-chlorotoluene and ethyl disulfide.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Odor Odorless
Quantity 5g
Linear Formula CH3SO3Na
Formula Weight 118.09
Percent Purity 98%
Chemical Name or Material Sodium methanesulfonate