Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium methoxide, ACS reagent, 0.5M solution in methanol, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
124-41-4,67-56-1
CH3NaO
54.02
MFCD00012179
WQDUMFSSJAZKTM-UHFFFAOYSA-N
sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
10942334
sodium;methanolate
C[O-].[Na+]
124-41-4,67-56-1
CH3NaO
54.02
MFCD00012179
WQDUMFSSJAZKTM-UHFFFAOYSA-N
sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
10942334
sodium;methanolate
C[O-].[Na+]
Linear Formula CH3ONa
Merck Index 15,8775
Solubility Information Solubility in water: reacts
Formula Weight 54.02
Grade ACS Reagent
Clarity Passes test
Density 0.8000g/mL
Packaging AcroSeal™ Glass bottle
Solubility Solubility in water: reacts.
Specific Gravity 0.8
Boiling Point 65.0°C
Melting Point -98.0°C
Quantity 800mL
Flash Point 11°C
Chemical Name or Material Sodium methoxide, 0.5M solution in methanol, AcroSeal™
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact

GHS Signal Word: Danger

EINECSNumber : 204-699-5