Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium methoxide, pure, anhydrous powder, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic Drum
124-41-4,497-19-8,1310-73-2
CH3NaO
54.02
MFCD00012179
WQDUMFSSJAZKTM-UHFFFAOYSA-N
sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
10942334
sodium;methanolate
C[O-].[Na+]
124-41-4,497-19-8,1310-73-2
CH3NaO
54.02
MFCD00012179
WQDUMFSSJAZKTM-UHFFFAOYSA-N
sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
10942334
sodium;methanolate
C[O-].[Na+]
Melting Point 126.0°C
Packaging Plastic Drum
Density 0.8
Color White-Yellow
Quantity 2.5kg
Alkalinity total alkalinity (as NaOCH3):> 99%
Linear Formula CH3ONa
UN Number 1431
Beilstein 01, 280
Fieser 01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352
Merck Index 15, 8775
Solubility Information Solubility in water: reacts with water. Other solubilities: soluble in methanol and ethanol, insoluble in hydrocarbons(benzene, toluene, xylene
Formula Weight 54.02
Physical Form Powder
Percent Purity ≥97% (NaOCH3)
Grade Pure
Chemical Name or Material Sodium methoxide
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Self-heating; may catch fire.
Flammable solid.
Harmful if swallowed.
May be corrosive to metals.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 204-699-5

RTECSNumber : PC3570000

TSCA : TSCA