Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Molybdate, Dihydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

10102-40-6
H4MoNa2O6
241.956
FDEIWTXVNPKYDL-UHFFFAOYSA-N
16211258
CHEBI:75213

High quality chemicals for laboratory and specialized industrial use

CAS 10102-40-6
Molecular Formula H4MoNa2O6
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
SMILES O.O.[O-][Mo](=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;dioxido(dioxo)molybdenum;dihydrate
Molecular Weight (g/mol) 241.956
PubChem CID 16211258
ChEBI CHEBI:75213
Formula Weight 241.95
Packaging Poly Bottle
Specific Gravity 1L = 3.28kg
Quantity 2.5kg
Chemical Name or Material Sodium Molybdate, Dihydrate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less