Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium molybdate(VI) dihydrate, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
10102-40-6
MoNa2O4·2H2O
241.95
MFCD00149170
FDEIWTXVNPKYDL-UHFFFAOYSA-N
sodium molybdate dihydrate,sodium molybdate vi dihydrate,disodium molybdate dihydrate,unii-8f2sxi1704,na2moo4.2h2o,disodium dihydrate molybdate,molybdic acid sodium salt dihydrate,molybdic acid, disodium salt, dihydrate,sodium molybdate 2 hydrate,dipotassium dihydrate molybdate
16211258
CHEBI:75213
disodium;dioxido(dioxo)molybdenum;dihydrate
O.O.[O-][Mo](=O)(=O)[O-].[Na+].[Na+]
10102-40-6
MoNa2O4·2H2O
241.95
MFCD00149170
FDEIWTXVNPKYDL-UHFFFAOYSA-N
sodium molybdate dihydrate,sodium molybdate vi dihydrate,disodium molybdate dihydrate,unii-8f2sxi1704,na2moo4.2h2o,disodium dihydrate molybdate,molybdic acid sodium salt dihydrate,molybdic acid, disodium salt, dihydrate,sodium molybdate 2 hydrate,dipotassium dihydrate molybdate
16211258
CHEBI:75213
disodium;dioxido(dioxo)molybdenum;dihydrate
O.O.[O-][Mo](=O)(=O)[O-].[Na+].[Na+]
Oxidation reagent
Assay Percent Range 99% min
Linear Formula Na2MoO4·2H2O
Fieser 15,294
Merck Index 15, 8776
Solubility Information Solubility in water: 560g/L (o°C). Other solubilities: 1150g/l (100°C)
Formula Weight 241.95
Physical Form Crystalline Powder
Percent Purity 99+%
Density 3.2800g/mL
Packaging Glass Bottle
Solubility Solubility in water: 560g/L (o°C). Other solubilities: 1150g/l (100°C)
Specific Gravity 3.28
Color White
Quantity 5g
Chemical Name or Material Sodium molybdate(VI) dihydrate, 99+%
Show More Show Less

RTECSNumber : QA5085000