Sodium molybdenum oxide dihydrate, ACS, 99.5 to 103.0%, Alfa Aesar™

Manufacturer: Alfa Aesar 1221436

Quantity 500g

 View more versions of this product

Catalog No. AA1221436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10102-40-6
    Physical Form Powder
    Assay Percent Range 99.5-103.0%
    Molecular Formula MoNa2O4·2H2O
    Formula Weight 241.95 (205.92 Anhydrous)
    MDL Number MFCD00149170
    Synonym Sodium molybdate dihydrate
    Chemical Name or Material Sodium molybdenum oxide dihydrate, For analysis ACS
    Grade ACS Grade

    Specifications

    Formula Weight 241.95 (205.92 Anhydrous)
    Quantity 500g
    Color Green
    Density 2.71g/cm3 at 20°C
    EINECS Number 231-551-7
    Physical Form Powder
    Assay Percent Range 99.5-103.0%
    Appearance White powder/flake/crystalline/beads
    MDL Number MFCD00149170
    CAS 10102-40-6
    Odor Odorless
    Molecular Formula MoNa2O4·2H2O
    Synonym Sodium molybdate dihydrate
    Chemical Name or Material Sodium molybdenum oxide dihydrate, For analysis ACS
    Grade ACS Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit