Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium n-heptyl sulfate, 99% (dry wt.), water <2%

Click to view available options
Quantity:
1g
5g
25g
18981-98-1
C7H16NaO4S
219.251
MFCD00051524
SLGMFWJQQBPOMZ-UHFFFAOYSA-N
heptyl sodium sulfate
131875435
heptyl hydrogen sulfate;sodium
CCCCCCCOS(=O)(=O)O.[Na]
18981-98-1
C7H16NaO4S
219.251
MFCD00051524
SLGMFWJQQBPOMZ-UHFFFAOYSA-N
heptyl sodium sulfate
131875435
heptyl hydrogen sulfate;sodium
CCCCCCCOS(=O)(=O)O.[Na]
Quantity 5g
Melting Point 199°C to 202°C (decomposition)
Linear Formula CH3(CH2)6OSO3Na
Beilstein 6018116
Formula Weight 218.24
Percent Purity 99%
Chemical Name or Material Sodium n-heptyl sulfate
Assay (dry wt.), water <2%

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA