Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Sodium n-heptyl sulfate, 99% (dry wt.), water <2%
Click to view available options
Quantity:
1g
5g
25g
Specifications
Quantity | 25g |
Melting Point | 199°C to 202°C (decomposition) |
Linear Formula | CH3(CH2)6OSO3Na |
Beilstein | 6018116 |
Formula Weight | 218.24 |
Percent Purity | 99% |
Chemical Name or Material | Sodium n-heptyl sulfate |
Assay | (dry wt.), water <2% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
Recommended Storage : Ambient temperatures