Alfa Aesar™ Sodium n-tridecyl sulfate, 99%, may cont. up to ca 2% water and residual solvent

Manufacturer: Alfa Aesar™ B2208914

Quantity 25g

 View more versions of this product

Catalog No. AAB2208914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3026-63-9
    Molecular Formula C13H27NaO4S
    Linear Formula CH3(CH2)12OSO3Na
    Formula Weight 302.4
    MDL Number MFCD00024993
    Synonym n-Tridecyl sulfate sodium salt
    Chemical Name or Material Sodium n-tridecyl sulfate

    Specifications

    Formula Weight 302.4
    Molecular Formula C13H27NaO4S
    Linear Formula CH3(CH2)12OSO3Na
    MDL Number MFCD00024993
    CAS 3026-63-9
    Synonym n-Tridecyl sulfate sodium salt
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 185° to 189°C
    Quantity 25g
    Chemical Name or Material Sodium n-tridecyl sulfate
    Assay 99%, may cont. up to ≈2% water and residual solvent

    Description

    Applications
    Sodium n-tridecyl sulfate acts as a foaming agent and a wetting agent. It is used as a cleaning agent and finds application in industrial and household cleaning products and personal care products. Further, it serves as an emulsifying agent in the transport of hydrocarbons. In addition to this, it acts as an anionic surfactant.

    Solubility
    Soluble in water.

    Notes
    Incompatible with bases and strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit