Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium nicotinate, 98%

Click to view available options
Quantity:
100g
500g
54-86-4
C6H4NNaO2
145.093
MFCD00064352
KFLRWGSAMLBHBV-UHFFFAOYSA-M
sodium nicotinate,nicotinic acid sodium salt,sodium pyridine-3-carboxylate,natriumnicotinat german,unii-3zx78b4q2l,natriumnicotinat,3-pyridinecarboxylic acid, sodium salt,nicotinic acid, sodium salt,3-pyridinecarboxylic acid, sodium salt 1:1,nicotinic acid sodium
23687810
sodium;pyridine-3-carboxylate
C1=CC(=CN=C1)C(=O)[O-].[Na+]
54-86-4
C6H4NNaO2
145.093
MFCD00064352
KFLRWGSAMLBHBV-UHFFFAOYSA-M
sodium nicotinate,nicotinic acid sodium salt,sodium pyridine-3-carboxylate,natriumnicotinat german,unii-3zx78b4q2l,natriumnicotinat,3-pyridinecarboxylic acid, sodium salt,nicotinic acid, sodium salt,3-pyridinecarboxylic acid, sodium salt 1:1,nicotinic acid sodium
23687810
sodium;pyridine-3-carboxylate
C1=CC(=CN=C1)C(=O)[O-].[Na+]
Melting Point >300°C
Quantity 100g
Formula Weight 145.1
Percent Purity 98%
Chemical Name or Material Sodium nicotinate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 200-215-1

RTECSNumber : QT2060000

TSCA : Yes

Recommended Storage : Ambient temperatures