Alfa Aesar™ Sodium nitrate, 98+%

Manufacturer: Alfa Aesar™ A123270C

Quantity 10,000g

 View more versions of this product

Catalog No. AAA123270C

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7631-99-4
    Molecular Formula NaNO3
    Formula Weight 84.99
    MDL Number MFCD00011119
    UN Number UN1498
    Merck Index 14,8647
    Chemical Name or Material Sodium nitrate

    Specifications

    Formula Weight 84.99
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P280-P301+P312-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula NaNO3
    CAS 7631-99-4
    UN Number UN1498
    Quantity 10,000g
    MDL Number MFCD00011119
    Merck Index 14,8647
    Health Hazard 2 GHS H Statement
    H272-H302
    May intensify fire
    oxidizer.
    Harmful if swallowed.
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: SODIUM NITRATE
    RTECS Number WC5600000
    Odor Odorless
    Melting Point ≈308°C
    Health Hazard 1 Danger
    Chemical Name or Material Sodium nitrate
    Assay ≥98%

    Description

    Applications
    Sodium nitrate is used in the production of fertilizers, nitric acid, pyrotechnics, smoke-bombs, glass and pottery enamels. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent. Further it finds use as food preservative and as a solid rocket propellant. It is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems.

    Solubility
    Freely soluble in water, ammonia and hydrazine. Soluble in alcohol.Slightly soluble in pyridine.

    Notes
    Sodium nitrate is hygroscopic in nature.

    GHS H Statement
    H272-H302
    May intensify fire
    oxidizer.
    Harmful if swallowed.

    GHS P Statement
    P221-P210-P220-P280-P301+P312-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit