Sodium nitrate, 99.999% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1087322

Quantity 100g

 View more versions of this product

Catalog No. AA1087322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7631-99-4
    Physical Form Powder
    Assay Percent Range 99.999% (metals basis)
    Molecular Formula NaNO3
    Formula Weight 84.99
    MDL Number MFCD00011119
    Chemical Name or Material Sodium nitrate

    Specifications

    Formula Weight 84.99
    Quantity 100g
    DOT Information DOT Class: 5.1, Packaging Group: III
    EINECS Number 231-554-3
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.999% (metals basis)
    Molecular Formula NaNO3
    Appearance White crystals
    MDL Number MFCD00011119
    CAS 7631-99-4
    Health Hazard 1 H272-H302: May intensify fire; oxidizer. Harmful if swallowed.
    Chemical Name or Material Sodium nitrate

    H272-H302: May intensify fire; oxidizer. Harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit