Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrate, BiotechGrade, 99-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
6 x 500 g
Packaging:
Poly Bottle
7631-99-4
NNaO3
84.99
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium nitrate
[Na+].[O-][N+]([O-])=O
7631-99-4
NNaO3
84.99
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium nitrate
[Na+].[O-][N+]([O-])=O

S1347, 7631-99-4, NaNO3

Spectrum™ Sodium Nitrate, BiotechGrade is a white solid and soluble in water. It is useful in production of glass, fertilizers, pyrotechnics, and food preservatives. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Grade Biotech
CAS Max % 1
Appearance Clear granules
Arsenic (As) 3e-006
Percent Purity 99.0 to 100.5%
Insoluble Matter 0.0001
Iron (Fe) 3e-006
Lead (Pb) 5 ppm
Packaging Poly Bottle
pH 5.5 to 8.0
Quantity 500 g
Show More Show Less