Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Nitrate, Technical, Spectrum™ Chemical
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Sodium Nitrate, Technical , is a white solid that is particularly soluble in water and in used in the production of fertilizers, gunpowder, glass, pottery enamels, solid rocket propellant and others. It is also notably used in the production of food as a food additive.
Specifications
CAS Max % | 1 |
Quantity | 12 kg |
UN Number | UN1498 |
Grade | Technical |
Packaging | Poly Pail |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction