Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium nitrite, 99%, extra pure, contains an anticaking reagent, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
10kg
Packaging:
Plastic Bucket
Plastic bottle
7632-00-0
NNaO2
68.995
MFCD00011118
LPXPTNMVRIOKMN-UHFFFAOYSA-M
sodium nitrite,nitrous acid, sodium salt,sodiumnitrite,erinitrit,filmerine,natrium nitrit,nitrito sodico,anti-rust,nitrite, sodium,diazotizing salts
23668193
CHEBI:78870
sodium;nitrite
N(=O)[O-].[Na+]
7632-00-0
NNaO2
68.995
MFCD00011118
LPXPTNMVRIOKMN-UHFFFAOYSA-M
sodium nitrite,nitrous acid, sodium salt,sodiumnitrite,erinitrit,filmerine,natrium nitrit,nitrito sodico,anti-rust,nitrite, sodium,diazotizing salts
23668193
CHEBI:78870
sodium;nitrite
N(=O)[O-].[Na+]
Additional Information anticaking reagent: < 0.1%
Assay Percent Range 99%
Linear Formula NaNO2
Fieser 01,1097; 02,386; 04,459; 06,547; 09,432; 11,491; 13,170; 16,86
Merck Index 15, 8779
Formula Weight 69
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Grade Extra Pure
Packaging Plastic bottle
Water 0.3% max.
Color Yellow
Quantity 2.5kg
Chemical Name or Material Sodium nitrite, 99%
Show More Show Less

GHS P Statement
Toxic if swallowed.
Very toxic to aquatic life.
Causes serious eye irritation.
May intensify fire; oxidizer.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 231-555-9

RTECSNumber : RA1225000

TSCA : TSCA