Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrite, For Analysis, EMSURE™, MilliporeSigma™

Click to view available options
Quantity:
100g
12kg
Packaging:
Plastic Bottle
Plastic Bucket
7632-00-0
NNaO2
69.00
MFCD00011118
LPXPTNMVRIOKMN-UHFFFAOYSA-M
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
23668193
CHEBI:78870
sodium nitrite
[Na+].[O-]N=O
7632-00-0
NNaO2
69.00
MFCD00011118
LPXPTNMVRIOKMN-UHFFFAOYSA-M
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
23668193
CHEBI:78870
sodium nitrite
[Na+].[O-]N=O

Recommended Storage

+5°C to +30°C.

Solubility Information 820g/L
Formula Weight 69
Physical Form Solid
Percent Purity >99.0%
Grade ACS
Density 2.1 g/3 (20 °C)
Packaging Plastic Bottle
Storage Information +5°C to +30°C.
Color White
Melting Point 280°C (decomposition)
pH 9 (100g/L, H2O, 20°C)
Quantity 100g
Chemical Name or Material Sodium Nitrite
Show More Show Less

EINECSNumber : 007-010-00-4

RTECSNumber : RA1225000