Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrite, Granular, FCC, 97-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7632-00-0
NNaO2
69.00
LPXPTNMVRIOKMN-UHFFFAOYSA-M
sodium nitrite
[Na+].[O-]N=O
7632-00-0
NNaO2
69.00
LPXPTNMVRIOKMN-UHFFFAOYSA-M
sodium nitrite
[Na+].[O-]N=O

SO194, 7632-00-0, NaNO2

Spectrum™ Sodium Nitrite, Granular, FCC is used as curing agent and food preservative that creates a distinct flavor, controls lipid oxidation, and acts as an antimicrobial. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Percent Purity 97.0 to 100.5%
Lead (Pb) 4 mg/kg
Loss on Drying 0.0025
Packaging Fiber Drum
Quantity 45 kg