Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrite, Granular, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7632-00-0
NNaO2
69.00
LPXPTNMVRIOKMN-UHFFFAOYSA-M
sodium nitrite
[Na+].[O-]N=O
7632-00-0
NNaO2
69.00
LPXPTNMVRIOKMN-UHFFFAOYSA-M
sodium nitrite
[Na+].[O-]N=O

SO185, 7632-00-0, NaNO2

Spectrum™ Sodium Nitrite, Granular, USP is used therapeutically as an antidote in cyanide poisoning. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Bacterial Endotoxins 0.1 USP
Percent Purity 98 to 102%
Grade USP
Carbonate 0.0002
Chloride 50 ppm
Insoluble Matter 50 ppm
Loss on Drying 0.0025
Packaging Poly Bottle
Sulfate 0.0001
pH 7.0 to 9.0
Quantity 500 g
Show More Show Less