Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitroferricyanide, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle

Chemical Identifiers

13755-38-9
C5H4FeN6Na2O3
297.951
XRKMNJXYOFSTBE-UHFFFAOYSA-N
11953895
disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[N-]=O.O.O.[Na+].[Na+].[Fe+4]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 297.95
Physical Form Crystals
Packaging Poly Bottle
Quantity 500g
Chemical Name or Material Sodium Nitroferricyanide, Dihydrate
Grade Baker Analyzed™ ACS Reagent