Alfa Aesar™ Sodium orthosilicate, (mixture of NaOH and Na2SiO3 yielding » Na4SiO4 in solution)

Manufacturer: Alfa Aesar™ 17568A7

Quantity 5kg

 View more versions of this product

Catalog No. AA17568A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13472-30-5
    MDL Number MFCD00014252
    UN Number UN1759
    Synonym Sodium silicate (ortho)
    Chemical Name or Material Sodium orthosilicate, Mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution

    Specifications

    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    UN Number UN1759
    Color White
    Quantity 5kg
    Health Hazard 2 GHS H Statement
    H314-H335
    Causes severe skin burns and eye damage.
    May cause respiratory irritation.
    Odor Odorless
    MDL Number MFCD00014252
    Synonym Sodium silicate (ortho)
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
    TSCA TSCA
    CAS 13472-30-5
    Chemical Name or Material Sodium orthosilicate, Mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution

    GHS H Statement
    H314-H335
    Causes severe skin burns and eye damage.
    May cause respiratory irritation.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit