Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium oxalate, 98.5%, extra pure, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
62-76-0
C2Na2O4
134
MFCD00012465
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
sodium oxalate,disodium oxalate,natriumoxalat,ethanedioic acid, disodium salt,oxalic acid, disodium salt,natriumoxalat german,stavelan sodny czech,oxalic acid disodium salt,unii-7u0v68lt9x,ethanedioic acid disodium salt
6125
disodium;oxalate
C(=O)(C(=O)[O-])[O-].[Na+].[Na+]
62-76-0
C2Na2O4
134
MFCD00012465
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
sodium oxalate,disodium oxalate,natriumoxalat,ethanedioic acid, disodium salt,oxalic acid, disodium salt,natriumoxalat german,stavelan sodny czech,oxalic acid disodium salt,unii-7u0v68lt9x,ethanedioic acid disodium salt
6125
disodium;oxalate
C(=O)(C(=O)[O-])[O-].[Na+].[Na+]
Insoluble Matter 0.2% max. (in water)
Color White
Melting Point 250.0°C to 270.0°C
Packaging Plastic bottle
Quantity 500g
Loss on Drying 0.1% max. (105°C, 2 hrs)
Assay Percent Range 98.5%
Linear Formula NaO2CCO2Na
Merck Index 15, 8781
Solubility Information Solubility in water: 37g/L (20°C). Other solubilities: 6.3g/100mL water (100°C),insoluble in ethanol and ether
Formula Weight 134
Physical Form Fine Powder
Percent Purity 98.5%
Grade Extra Pure
Chemical Name or Material Sodium oxalate, pure
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-550-3

RTECSNumber : KI1750000

TSCA : TSCA