Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium oxalate, ACS reagent, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
62-76-0
C2Na2O4
134
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
sodium oxalate,disodium oxalate,natriumoxalat,ethanedioic acid, disodium salt,oxalic acid, disodium salt,natriumoxalat german,stavelan sodny czech,oxalic acid disodium salt,unii-7u0v68lt9x,ethanedioic acid disodium salt
6125
disodium;oxalate
C(=O)(C(=O)[O-])[O-].[Na+].[Na+]
62-76-0
C2Na2O4
134
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
sodium oxalate,disodium oxalate,natriumoxalat,ethanedioic acid, disodium salt,oxalic acid, disodium salt,natriumoxalat german,stavelan sodny czech,oxalic acid disodium salt,unii-7u0v68lt9x,ethanedioic acid disodium salt
6125
disodium;oxalate
C(=O)(C(=O)[O-])[O-].[Na+].[Na+]
Linear Formula NaO2CCO2Na
Ammonium 0.002% max
Merck Index 15,8781
Solubility Information Solubility in water: 37g/L (20°C). Other solubilities: 6.3g/100mL water (100°C),insoluble in ethanol and ether
Formula Weight 134
Grade ACS Reagent
Chloride 0.002% max
Insoluble Matter 0.005% max
Packaging Plastic bottle
Solubility Solubility in water: 37g/L (20°C). Other solubilities: 6.3 g/100mL water (100°C), insoluble in ethanol and ether
Sulfate 0.002% max
Melting Point 250.0°C to 270.0°C
Quantity 100g
Chemical Name or Material Sodium oxalate
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 200-550-3