Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Oxalate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

62-76-0
C2Na2O4
133.998
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
6125

High quality chemicals for laboratory and specialized industrial use

CAS 62-76-0
Molecular Formula C2Na2O4
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
SMILES C(=O)(C(=O)[O-])[O-].[Na+].[Na+]
IUPAC Name disodium;oxalate
Molecular Weight (g/mol) 133.998
PubChem CID 6125
Formula Weight 134
Physical Form Powder
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.34kg
Quantity 125g
Chemical Name or Material Sodium Oxalate, Titrimetry Standard
Grade Baker Analyzed™ ACS Reagent
Show More Show Less