Learn More
Sodium pentacyanonitrosylferrate(III) dihydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals
Description
For chromatographic detection of peptides.Sodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Red |
Physical Form | Crystals |
Quantity | 50 g |
Assay Percent Range | 99.0-102.0% |
Solubility Information | Soluble in water. Slightly soluble ethanol. |
Formula Weight | 297.95 (261.92 Anhydrous) |
Grade | ACS |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 1.72 |
Show More |
Safety and Handling
Hazard Category | H301 |
Precautionary Statement | P260-P264b-P270-P271-P301+P310-P304+P340-P312-P314-P330-P501c |
DOTInformation | DOT Class: 6.1, Packaging Group: III |
EINECSNumber | 238-373-9 |
RTECSNumber | LJ8925000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.