Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium pentacyanonitrosylferrate(III) dihydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals

Catalog Number AA8768318
Click to view available options
Quantity:
50 g
250 g
13755-38-9
C5H4FeN6Na2O3
297.95
MFCD00149192
JFPDVRGGFBUORC-UHFFFAOYSA-N
nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
11953895
disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
13755-38-9
C5H4FeN6Na2O3
297.95
MFCD00149192
JFPDVRGGFBUORC-UHFFFAOYSA-N
nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
11953895
disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

For Chromatographic Detection of Peptides

For chromatographic detection of peptides.Sodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Red
Physical Form Crystals
Quantity 50 g
Assay Percent Range 99.0-102.0%
Solubility Information Soluble in water. Slightly soluble ethanol.
Formula Weight 297.95 (261.92 Anhydrous)
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Density 1.72
Chemical Name or Material Sodium pentacyanonitrosylferrate(III) dihydrate
Show More Show Less
Hazard Category H301
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P312-P314-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 238-373-9
RTECSNumber LJ8925000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit