Alfa Aesar™ Sodium perborate tetrahydrate, 97%

Manufacturer: Alfa Aesar™ A1018922

Quantity 100g

 View more versions of this product

Catalog No. AAA1018922

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 10486-00-7
    Molecular Formula NaBO3·4H2O
    Formula Weight 153.86 (81.84 Anhydrous)
    MDL Number MFCD00149231
    UN Number UN1479
    Merck Index 14,8652
    Chemical Name or Material Sodium perborate tetrahydrate

    Specifications

    Formula Weight 153.86 (81.84 Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1479
    Health Hazard 2 GHS H Statement
    H272-H360-H318-H302-H335
    May intensify fire
    oxidizer.
    May damage fertility or the unborn child.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
    CAS 10486-00-7
    TSCA TSCA
    Molecular Formula NaBO3·4H2O
    MDL Number MFCD00149231
    Merck Index 14,8652
    RTECS Number SC7350000
    Health Hazard 1 Danger
    Melting Point 60°C (dec.)
    pH ≈10
    Quantity 100g
    Odor Odorless
    Chemical Name or Material Sodium perborate tetrahydrate
    Assay 97%

    Description

    Applications
    Sodium perborate tetrahydrate is considered as a source of active hydrogen used in detergents, cleaning products, laundry detergents and bleaches. It finds application as a tooth bleaching agent utilized for tooth. It has antiseptic properties and acts as disinfectant as well as preservative in eye drops. Further, it is mixed with suitable activator, tetraacetylethylenediamine in order to release oxygen at lower temperatures.

    Solubility
    Soluble in water.

    Notes
    Moisture sensitive.Incompatible with metals, metallic salts, acids, bases and reducing agents.

    GHS H Statement
    H272-H360-H318-H302-H335
    May intensify fire
    oxidizer.
    May damage fertility or the unborn child.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit