Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium perborate tetrahydrate, 97%

Manufacturer:  Alfa Aesar™ A1018936

Quantity 500g

 View more versions of this product

Catalog No. AAA1018936


  Check Availability
Add to cart

Specifications

Description

Applications
Sodium perborate tetrahydrate is considered as a source of active hydrogen used in detergents, cleaning products, laundry detergents and bleaches. It finds application as a tooth bleaching agent utilized for tooth. It has antiseptic properties and acts as disinfectant as well as preservative in eye drops. Further, it is mixed with suitable activator, tetraacetylethylenediamine in order to release oxygen at lower temperatures.

Solubility
Soluble in water.

Notes
Moisture sensitive.Incompatible with metals, metallic salts, acids, bases and reducing agents.
Description & Specifications

Chemical Identifiers

10486-00-7
153.86 (81.84 Anhydrous)
UN1479
Sodium perborate tetrahydrate
NaBO3·4H2O
MFCD00149231
14,8652

Specifications

10486-00-7
153.86 (81.84 Anhydrous)
UN1479
60°C (dec.)
500g
Sodium perborate tetrahydrate
NaBO3·4H2O
MFCD00149231
14,8652
≈10
Odorless
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H272-H360-H318-H302-H335
May intensify fire
oxidizer.
May damage fertility or the unborn child.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

RTECSNumber : SC7350000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit