Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium perchlorate, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic bottle
7601-89-0
ClNaO4
122.44
BAZAXWOYCMUHIX-UHFFFAOYSA-M
sodium perchlorate,perchloric acid, sodium salt,natriumperchlorat,perchlorate de sodium,natriumperchloraat dutch,natriumperchlorat german,unii-97f4mty3va,cpl 46,perchlorate de sodium french,sodio perclorato di italian
522606
sodium;perchlorate
[O-]Cl(=O)(=O)=O.[Na+]
7601-89-0
ClNaO4
122.44
BAZAXWOYCMUHIX-UHFFFAOYSA-M
sodium perchlorate,perchloric acid, sodium salt,natriumperchlorat,perchlorate de sodium,natriumperchloraat dutch,natriumperchlorat german,unii-97f4mty3va,cpl 46,perchlorate de sodium french,sodio perclorato di italian
522606
sodium;perchlorate
[O-]Cl(=O)(=O)=O.[Na+]
Oxidation reagent
Assay Percent Range 99% min. (on dry substance)
Linear Formula NaClO4
Merck Index 15,8784
Solubility Information Solubility in water: 209g/100mL (15°C). Other solubilities: 170g/100mL water (0°C),300g/100mL water (80°C),soluble in alcohol
Formula Weight 122.44
Physical Form Crystals or Crystalline Powder
Percent Purity 99+%
Grade Analysis
Calcium (Ca) 200ppm max.
Chloride 30ppm max
Chlorine (Cl) 30ppm max.
Heavy Metals (as Pb) 5ppm max.
Insoluble in Water 0.005% max.
Insoluble Matter 0.005% max. (in water)
Iron (Fe) 5ppm max.
Loss on Drying 2.0% max. (130°C, 2 hrs)
Packaging Plastic bottle
Potassium (K) 500ppm max.
Solubility Solubility in water: 209 g/100mL (15°C). Other solubilities: 170 g/100mL water (0°C), 300 g/100mL water (80°C), soluble in alcohol
Sulfate 20ppm max.
Color White
Melting Point 482.0°C
pH 6.0 to 8.0 (5% aq. soln.)
Quantity 250g
Chemical Name or Material Sodium perchlorate
Show More Show Less

GHS H Statement
May cause fire or explosion; strong oxidizer.
Harmful if swallowed.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.

GHS Signal Word: Danger

EINECSNumber : 231-511-9