Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium perchlorate, anhydrous, ACS, 98.0 to 102.0%, Alfa Aesar™

Click to view available options
Quantity:
100g
500g
7601-89-0
ClNaO4
122.436
MFCD00003480
BAZAXWOYCMUHIX-UHFFFAOYSA-M
sodium perchlorate,perchloric acid, sodium salt,natriumperchlorat,perchlorate de sodium,natriumperchloraat dutch,natriumperchlorat german,unii-97f4mty3va,cpl 46,perchlorate de sodium french,sodio perclorato di italian
522606
sodium;perchlorate
[O-]Cl(=O)(=O)=O.[Na+]
7601-89-0
ClNaO4
122.436
MFCD00003480
BAZAXWOYCMUHIX-UHFFFAOYSA-M
sodium perchlorate,perchloric acid, sodium salt,natriumperchlorat,perchlorate de sodium,natriumperchloraat dutch,natriumperchlorat german,unii-97f4mty3va,cpl 46,perchlorate de sodium french,sodio perclorato di italian
522606
sodium;perchlorate
[O-]Cl(=O)(=O)=O.[Na+]
Assay Percent Range 98.0 to 102.0%
Solubility Information Soluble in water and alcohol.
Formula Weight 122.46
Physical Form Crystalline
Grade ACS Reagent
Density 2.50 g/mL
Sensitivity Hygroscopic
Melting Point 482°C (decomposition)
Quantity 500g
Chemical Name or Material Sodium perchlorate, anhydrous, For analysis ACS

P210-P220-P221-P260-P264b-P270-P280-P283-P301+P312-P305+P351+P338-P306+P360-P314-P330-P370+P378q-P371+P380+P375-P501c

H271-H302-H319-H373

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 231-511-9

TSCA : Yes

Recommended Storage : Ambient temperatures