Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium perchlorate monohydrate, 85+%, ACS reagent, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
2.5kg
Packaging:
Plastic bottle
7791-07-3
ClNaO4·H2O
140.46
IXGNPUSUVRTQGW-UHFFFAOYSA-M
sodium perchlorate monohydrate,sodium perchlorate hydrate,unii-g4h485lhcw,g4h485lhcw,ccris 8893,perchloric acid, sodium salt, monohydrate,sodium hydrate perchlorate,clo4.na.h2o,acmc-1bii1,sodium perchlorate-monohydrate
516899
sodium;perchlorate;hydrate
O.[O-]Cl(=O)(=O)=O.[Na+]
7791-07-3
ClNaO4·H2O
140.46
IXGNPUSUVRTQGW-UHFFFAOYSA-M
sodium perchlorate monohydrate,sodium perchlorate hydrate,unii-g4h485lhcw,g4h485lhcw,ccris 8893,perchloric acid, sodium salt, monohydrate,sodium hydrate perchlorate,clo4.na.h2o,acmc-1bii1,sodium perchlorate-monohydrate
516899
sodium;perchlorate;hydrate
O.[O-]Cl(=O)(=O)=O.[Na+]
Oxidation reagent
Assay Percent Range 85.0 to 90% (as NaClO4) (Argentometry)
Linear Formula ClNaO4·H2O
Merck Index 15,8784
Solubility Information Solubility in water: 2090g/L (15°C). Other solubilities: soluble in alcohol
Formula Weight 140.46
Physical Form Crystalline Powder
Percent Purity 85+%
Grade ACS Reagent
Calcium (Ca) 200ppm max.
Chloride 30ppm max.
Chlorine (Cl) 30ppm max.
Heavy Metals (as Pb) 5ppm max.
Insoluble in Water 0.005% max.
Insoluble Matter 0.005% max
Iron (Fe) 5ppm max.
Packaging Plastic bottle
Potassium (K) 500ppm max.
Solubility Solubility in water: 2090g/L (15°C). Other solubilities: soluble in alcohol
Sulfate 20ppm max.
Color White to Yellow
Melting Point 130.0°C
pH 6.0 to 8.0 (5% aq. soln. at 25°C)
Quantity 2.5kg
Chemical Name or Material Sodium perchlorate monohydrate
Show More Show Less

GHS H Statement
May cause fire or explosion; strong oxidizer.
Harmful if swallowed.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.

GHS Signal Word: Danger