Alfa Aesar™ Sodium perchlorate monohydrate, 97+%

Manufacturer: Alfa Aesar™ A1124330

Quantity 250g

 View more versions of this product

Catalog No. AAA1124330

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7791-07-3
    Molecular Formula NaClO4·H2O
    Formula Weight 140.48 (122.46 Anhydrous)
    MDL Number MFCD00149164
    UN Number UN1502
    Merck Index 14,8653
    Chemical Name or Material Sodium perchlorate monohydrate

    Specifications

    Formula Weight 140.48 (122.46 Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P283-P210-P306+P360-P371+P380+P375-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
    In case of major fire and large quantities: Evacuate area.
    Fight fire remotely due to the risk of explosion.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H271-H302
    May cause fire or explosion
    strong oxidizer.
    Harmful if swallowed.
    CAS 7791-07-3
    UN Number UN1502
    Melting Point 130°C (dec.)
    Quantity 250g
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: SODIUM PERCHLORATE
    Odor Odorless
    MDL Number MFCD00149164
    Merck Index 14,8653
    Health Hazard 1 Danger
    RTECS Number SC9850000
    TSCA TSCA
    Molecular Formula NaClO4·H2O
    Chemical Name or Material Sodium perchlorate monohydrate
    Assay ≥97%

    Description

    Applications
    Sodium perchlorate, monohydrate is a chaotropic agent used in standard DNA extraction and hybridization reactions in molecular biology. It is widely utilized as a deproteinization agent during nucleic acid synthesis. A bit of modern commercial explosives products may contain sodium perchlorate, ammonium perchlorate, or potassium perchlorate as a chemical sensitizing agent.

    Solubility
    Soluble in water.

    Notes
    It is a hygroscopic solid. Store under inert atmosphere and keep away from ignition sources.

    GHS H Statement
    H271-H302
    May cause fire or explosion
    strong oxidizer.
    Harmful if swallowed.

    GHS P Statement
    P221-P283-P210-P306+P360-P371+P380+P375-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
    In case of major fire and large quantities: Evacuate area.
    Fight fire remotely due to the risk of explosion.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit