Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Sodium Periodate, Honeywell Fluka™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
7790-28-5
INaO4
213.89
MFCD00003534
JQWHASGSAFIOCM-UHFFFAOYSA-M
sodium periodate,sodium metaperiodate,sodium m-periodate,sodiumperiodate,sodium meta periodate,periodic acid, sodium salt,sodium meta-periodate,periodic acid hio4 , sodium salt,periodate sodium,sodium penodate
23667635
CHEBI:75226
sodium;periodate
[O-]I(=O)(=O)=O.[Na+]
7790-28-5
INaO4
213.89
MFCD00003534
JQWHASGSAFIOCM-UHFFFAOYSA-M
sodium periodate,sodium metaperiodate,sodium m-periodate,sodiumperiodate,sodium meta periodate,periodic acid, sodium salt,sodium meta-periodate,periodic acid hio4 , sodium salt,periodate sodium,sodium penodate
23667635
CHEBI:75226
sodium;periodate
[O-]I(=O)(=O)=O.[Na+]

ACS Reagent,≥99.8%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula NaIO4
UN Number 3085
Formula Weight 213.89g/mol
Percent Purity ≥99.8%
Grade ACS Reagent
Quantity 2.5kg
Chemical Name or Material Sodium Periodate

P220-P273-P280-P305 + P351 + P338-P310

H271-H314-H372-H400

EINECSNumber : 232-197-6

RTECSNumber : SD4550000