Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Peroxide, Reagent, ACS, 93%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
Steel Can
1313-60-6
Na2O2
77.98
PFUVRDFDKPNGAV-UHFFFAOYSA-N
disodium dioxidanediide
[Na+].[Na+].[O-][O-]
1313-60-6
Na2O2
77.98
PFUVRDFDKPNGAV-UHFFFAOYSA-N
disodium dioxidanediide
[Na+].[Na+].[O-][O-]

S1390, 1313-60-6, Na2O2

Spectrum™ Sodium Peroxide, Reagent, ACS is a solid that comes in the form yellow micro beads. It is an inorganic compound that was historically used to bleach wood pulp in order to prepare it for the use in paper manufacture. Today it is most commonly used for certain specialized lab operations such as mineral extraction form ore. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Grade ACS
CAS Max % 1
Percent Purity 93%
Heavy Metals (as Pb) 2e-005
Iron (Fe) 5e-005
Packaging Steel Can
Sulfate 1e-005
Quantity 125 g