Sodium perrhenate, 99.95% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 6210703

Quantity 1g

 View more versions of this product

Catalog No. AA6210703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13472-33-8
    Molecular Formula NaO4Re
    Linear Formula NaReO4
    Formula Weight 273.19
    MDL Number MFCD00014231
    UN Number UN1479
    Chemical Name or Material Sodium perrhenate

    Specifications

    Formula Weight 273.19
    Quantity 1g
    CAS 13472-33-8
    Color White
    Melting Point 300°C
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P280-P305+P351+P338-P501a;
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
    TSCA TSCA
    Odor Odorless
    Physical Form Crystalline
    Sensitivity Hygroscopic
    Molecular Formula NaO4Re
    Linear Formula NaReO4
    MDL Number MFCD00014231
    UN Number UN1479
    Health Hazard 2 GHS H Statement
    H272-H315-H319;
    May intensify fire; oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    Density 5.390
    RTECS Number WD3675000
    Chemical Name or Material Sodium perrhenate
    Assay 99.95%, (metals basis)

    GHS H Statement
    H272-H315-H319;
    May intensify fire; oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P221-P210-P220-P280-P305+P351+P338-P501a;
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit