Sodium perrhenate, 99.95% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 6210701

Quantity 0.25g

 View more versions of this product

Catalog No. AA6210701

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-28-9
    Physical Form Powder
    Assay Percent Range 99.9% (metals basis)
    Molecular Formula ReO3
    Formula Weight 236.2
    MDL Number MFCD00016306
    Chemical Name or Material Rhenium(VI) oxide

    Specifications

    Physical Form Powder
    Formula Weight 236.2
    Quantity 0.25g
    Color Violet
    EINECS Number 215-228-8
    Assay Percent Range 99.9% (metals basis)
    MDL Number MFCD00016306
    Odor Odorless
    Molecular Formula ReO3
    CAS 1314-28-9
    Chemical Name or Material Rhenium(VI) oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit