Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium phenoxide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Sodium phenoxide is used as a precursor to aryl ethers and involved in the preparation of phenyl ethers and metal phenolates. It serves as an antiseptic and used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium phenoxide is used as a precursor to aryl ethers and involved in the preparation of phenyl ethers and metal phenolates. It serves as an antiseptic and used in organic synthesis.
Solubility
Very soluble in water, soluble in alcoholSoluble in water, acetone and alcohol.
Notes
Hygroscopic and air sensitive. Incompatible with oxidizing agents.
Sodium phenoxide is used as a precursor to aryl ethers and involved in the preparation of phenyl ethers and metal phenolates. It serves as an antiseptic and used in organic synthesis.
Solubility
Very soluble in water, soluble in alcoholSoluble in water, acetone and alcohol.
Notes
Hygroscopic and air sensitive. Incompatible with oxidizing agents.
Specifications
Melting Point | >300°C |
Odor | Phenol-like |
Linear Formula | C6H5ONa |
Quantity | 50 g |
UN Number | UN2905 |
Sensitivity | Air Sensitive |
Solubility Information | Very soluble in water,soluble in alcoholSoluble in water,acetone and alcohol. |
IUPAC Name | sodium;phenoxide |
Formula Weight | 116.09 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | GHS H Statement H314-H318-H302 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: PHENOLATES, SOLID |
EINECSNumber | 205-347-3 |
RTECSNumber | SM8780000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction