Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium phosphate, 0.2M buffer soln., pH 8.0, low endotoxin

Click to view available options
Quantity:
100mL
250mL
7732-18-5
MFCD00146206
7732-18-5
MFCD00146206
Concentration 0.2M buffer soln.
Physical Form Liquid
pH 8.0
Color Colorless
Quantity 250mL
Chemical Name or Material Sodium phosphate, low endotoxin