Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate, Dibasic, 7-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7782-85-6
H15Na2O11P
268.062
PYLIXCKOHOHGKQ-UHFFFAOYSA-L
6096963

High quality chemicals for laboratory and specialized industrial use

CAS 7782-85-6
Molecular Formula H15Na2O11P
InChI Key PYLIXCKOHOHGKQ-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]
IUPAC Name disodium;hydrogen phosphate;heptahydrate
Molecular Weight (g/mol) 268.062
PubChem CID 6096963
Formula Weight 268.07
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.70kg
Quantity 500g
Chemical Name or Material Sodium Phosphate, Dibasic, 7-Hydrate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less